CAS RN 321438-86-2
IUPAC Name:
[6-(methylsulfanyl)pyridin-3-yl]boronic acid
Synonyms:
6-(methylsulfanyl)pyridin-3-ylboronic acid
Molecular Weight (g/mol):
169.01
Molecular Formula:
C6H8BNO2S
InChi Key:
UXWKVPJOPVIIRU-UHFFFAOYSA-N
SMILES:
CSC1=CC=C(C=N1)B(O)O