accessibility menu, dialog, popup

UserName

HHClClNHNOO

CAS RN 32231-06-4

IUPAC Name: dihydrogen 1-[(2H-1,3-benzodioxol-5-yl)methyl]piperazine dichloride
Synonyms: methylenedioxybenzylpiperazine
Molecular Weight (g/mol): 293.19
Molecular Formula: C12H18Cl2N2O2
InChi Key: GLQHKFNNJILFTF-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].C(N1CCNCC1)C1=CC=C2OCOC2=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results