accessibility menu, dialog, popup

UserName

CAS RN 32459-62-4

IUPAC Name: 1-ethoxy-4-isocyanatobenzene
Synonyms: 1-ethoxy-4-isocyanatobenzene
Molecular Weight (g/mol): 163.18
Molecular Formula: C9H9NO2
InChi Key: FMYVTFRADSNGDN-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(C=C1)N=C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results