accessibility menu, dialog, popup

UserName

H3COOOCH3

CAS RN 3249-68-1

IUPAC Name: ethyl 3-oxohexanoate
Synonyms: ethyl 3-oxohexanoate
Molecular Weight (g/mol): 158.2
Molecular Formula: C8H14O3
InChi Key: KQWWVLVLVYYYDT-UHFFFAOYSA-N
SMILES: CCCC(=O)CC(=O)OCC
Boiling Point 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)