CAS RN 3257-18-9
CAS RN 3257-18-9
IUPAC Name:
2-{[(benzyloxy)carbonyl]amino}-3-(benzylsulfanyl)propanoic acid
Synonyms:
2-{[(benzyloxy)carbonyl]amino}-3-(benzylsulfanyl)propanoic acid
Molecular Weight (g/mol):
345.41
Molecular Formula:
C18H19NO4S
InChi Key:
ATPNWHGYKFXQNF-UHFFFAOYNA-N
SMILES:
OC(=O)C(CSCC1=CC=CC=C1)NC(=O)OCC1=CC=CC=C1
1
–
1
of
1
results
1
Z-Cys(Bzl)-OH, TRC
CAS: 3257-18-9 Molecular Formula: C18H19NO4S Molecular Weight (g/mol): 345.41 IUPAC Name: (2R)-3-benzylsulfanyl-2-(phenylmethoxycarbonylamino)propanoic acid SMILES: OC(=O)[C@H](CSCc1ccccc1)NC(=O)OCc2ccccc2