accessibility menu, dialog, popup

UserName

CAS RN 327-56-0

H3C(R)NH2OHO

CAS RN 327-56-0

IUPAC Name: 2-aminohexanoic acid
Synonyms: norleucine L-aminohexanoic acid (S)-aminohexanoic acid (S)-aminohexanoate
Molecular Weight (g/mol): 131.18
Molecular Formula: C6H13NO2
InChi Key: LRQKBLKVPFOOQJ-UHFFFAOYNA-N
SMILES: CCCCC(N)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)