accessibility menu, dialog, popup

UserName

NaOPOOONa

CAS RN 3279-54-7

IUPAC Name: phenoxyphosphonic acid sodium
Synonyms: phenylphosphate sodium
Molecular Weight (g/mol): 197.08
Molecular Formula: C6H7NaO4P
InChi Key: YXDQIUYZDWYOQN-UHFFFAOYSA-N
SMILES: [Na].OP(O)(=O)OC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results