accessibility menu, dialog, popup

UserName

CAS RN 3279-90-1

IUPAC Name: 6-bromo-1,2,3,4-tetrahydroquinolin-2-one
Synonyms: 6-bromo-3,4-dihydro-1H-quinolin-2-one
Molecular Weight (g/mol): 226.07
Molecular Formula: C9H8BrNO
InChi Key: MQWZSSIUHXNNTM-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(NC(=O)CC2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results