accessibility menu, dialog, popup

UserName

CAS RN 328-38-1

H3CCH3(R)NH2OHO

CAS RN 328-38-1

IUPAC Name: (2R)-2-amino-4-methylpentanoic acid
Synonyms: D-leucine L isomer leucine leucine, L-isomer L-α-aminoisocaproate L-isomer leucine L-a-aminoisocaproic acid D-leucin leucina leucine,d L-(+)-leucine
Molecular Weight (g/mol): 131.18
Molecular Formula: C6H13NO2
InChi Key: ROHFNLRQFUQHCH-RXMQYKEDSA-N
SMILES: CC(C)C[C@@H](N)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (5)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)