accessibility menu, dialog, popup

UserName

NONBr

CAS RN 32863-31-3

IUPAC Name: 5-(bromomethyl)-2,1,3-benzoxadiazole
Synonyms: 5-(bromomethyl)-2,1,3-benzoxadiazole
Molecular Weight (g/mol): 213.03
Molecular Formula: C7H5BrN2O
InChi Key: BUJFLTNYWUEROF-UHFFFAOYSA-N
SMILES: BrCC1=CC2=NON=C2C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results