accessibility menu, dialog, popup

UserName

H3COOHNNH2

CAS RN 3290-99-1

IUPAC Name: 4-methoxybenzohydrazide
Synonyms: benzoylhydrazine, P-methoxy
Molecular Weight (g/mol): 166.18
Molecular Formula: C8H10N2O2
InChi Key: REKQLYUAUXYJSZ-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(=O)NN
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results