CAS RN 33050-32-7
CAS RN 33050-32-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
6-Chloro[1,2,4]triazolo[4,3-b]pyridazin-3(2H)-one, TRC
CAS: 33050-32-7 Molecular Formula: C5H3N4OCl Molecular Weight (g/mol): 170.56 IUPAC Name: 6-chloro-2H-[1,2,4]triazolo[4,3-b]pyridazin-3-one SMILES: Oc1nnc2ccc(Cl)nn12