accessibility menu, dialog, popup

UserName

CAS RN 33114-15-7

IUPAC Name: niobium(5+) cyclopenta-1,3-dien-1-ide tetrachloride
Synonyms: niobium(5+) cyclopenta-1,3-dien-1-ide tetrachloride
Molecular Weight (g/mol): 299.8
Molecular Formula: C5H5Cl4Nb
InChi Key: KAWQZSRKEBAOSY-UHFFFAOYSA-J
SMILES: [Cl-].[Cl-].[Cl-].[Cl-].[Nb+5].C1C=CC=[C-]1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results