accessibility menu, dialog, popup

UserName

CAS RN 331730-08-6

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-3-(furan-2-yl)propanoic acid; N-cyclohexylcyclohexanamine
Synonyms: 2-[(tert-butoxycarbonyl)amino]-3-(furan-2-yl)propanoic acid; dicha
Molecular Weight (g/mol): 436.59
Molecular Formula: C24H40N2O5
InChi Key: QYEOQPFAYJEDFE-UHFFFAOYNA-N
SMILES: C1CCC(CC1)NC1CCCCC1.CC(C)(C)OC(=O)NC(CC1=CC=CO1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results