CAS RN 3319-99-1
IUPAC Name:
2-(thiophen-2-yl)pyridine
Synonyms:
2-(thiophen-2-yl)pyridine
Molecular Weight (g/mol):
161.22
Molecular Formula:
C9H7NS
InChi Key:
QLPKTAFPRRIFQX-UHFFFAOYSA-N
SMILES:
S1C=CC=C1C1=CC=CC=N1
1
–
1
of
1
results
1
2-(2-Thienyl)pyridine, 97%
CAS: 3319-99-1 Molecular Formula: C9H7NS Molecular Weight (g/mol): 161.22 MDL Number: MFCD00044441 InChI Key: QLPKTAFPRRIFQX-UHFFFAOYSA-N Synonym: 2-2-thienyl pyridine,2-thiophen-2-yl pyridine,2-thien-2-ylpyridine,2-2-pyridyl thiophene,pyridine, 2-2-thienyl,thienyl pyridine,2-thienylpyridine,2-pyrid-2-ylthiophene,acmc-1cknd,2-thien-2-yl pyridine PubChem CID: 76832 IUPAC Name: 2-thiophen-2-ylpyridine SMILES: S1C=CC=C1C1=CC=CC=N1