accessibility menu, dialog, popup

UserName

H3CONHOFFF

CAS RN 332-34-3

IUPAC Name: 2,2,2-trifluoro-N-(4-methoxyphenyl)acetamide
Synonyms: 2,2,2-trifluoro-N-(4-methoxyphenyl)acetamide
Molecular Weight (g/mol): 219.16
Molecular Formula: C9H8F3NO2
InChi Key: OUTSPSLYNAJENA-UHFFFAOYSA-N
SMILES: COC1=CC=C(NC(=O)C(F)(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results