accessibility menu, dialog, popup

UserName

CAS RN 332168-85-1

IUPAC Name: N-(3,5-dimethylphenyl)-2-fluorobenzamide
Synonyms: N-(3,5-dimethylphenyl)-2-fluorobenzamide
Molecular Weight (g/mol): 243.28
Molecular Formula: C15H14FNO
InChi Key: GKEBWXLAFINVJG-UHFFFAOYSA-N
SMILES: CC1=CC(NC(=O)C2=CC=CC=C2F)=CC(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results