accessibility menu, dialog, popup

UserName

CAS RN 3326-34-9

NH2OOHOOOH

CAS RN 3326-34-9

IUPAC Name: 5-amino-3',6'-dihydroxy-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
Synonyms: 5-amino-3',6'-dihydroxyspiro[2-benzofuran-1,9'-xanthen]-3-one
Molecular Weight (g/mol): 347.33
Molecular Formula: C20H13NO5
InChi Key: GZAJOEGTZDUSKS-UHFFFAOYSA-N
SMILES: NC1=CC=C2C(=C1)C(=O)OC21C2=CC=C(O)C=C2OC2=CC(O)=CC=C12
Color 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)