accessibility menu, dialog, popup

UserName

CAS RN 33371-97-0

CH3O(S)O

CAS RN 33371-97-0

IUPAC Name: (2S)-2-ethoxycyclohexan-1-one
Synonyms: (2S)-2-ethoxycyclohexan-1-one
Molecular Weight (g/mol): 142.2
Molecular Formula: C8H14O2
InChi Key: WBPWDGRYHFQTRC-QMMMGPOBSA-N
SMILES: CCO[C@H]1CCCCC1=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)