accessibility menu, dialog, popup

UserName

CAS RN 3343-39-3

HHClClNH2N

CAS RN 3343-39-3

IUPAC Name: dihydrogen 2-(pyridin-2-yl)ethan-1-amine dichloride
Synonyms: dihydrogen 2-pyridylethylamine dichloride
Molecular Weight (g/mol): 195.09
Molecular Formula: C7H12Cl2N2
InChi Key: GPIFKSYYILNJNR-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].NCCC1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results