accessibility menu, dialog, popup

UserName

CAS RN 3350-13-8

IUPAC Name: 2-{[(benzyloxy)carbonyl]amino}-4-{[(tert-butoxy)carbonyl]amino}butanoic acid; N-cyclohexylcyclohexanamine
Synonyms: 2-{[(benzyloxy)carbonyl]amino}-4-[(tert-butoxycarbonyl)amino]butanoic acid; dicha
Molecular Weight (g/mol): 533.71
Molecular Formula: C29H47N3O6
InChi Key: CYMIEBREXUYHGN-UHFFFAOYNA-N
SMILES: C1CCC(CC1)NC1CCCCC1.CC(C)(C)OC(=O)NCCC(NC(=O)OCC1=CC=CC=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results