accessibility menu, dialog, popup

UserName

CAS RN 33543-78-1

H3COONHN

CAS RN 33543-78-1

IUPAC Name: ethyl 1H-imidazole-2-carboxylate
Synonyms: ethyl 1H-imidazole-2-carboxylate
Molecular Weight (g/mol): 140.14
Molecular Formula: C6H8N2O2
InChi Key: UHYNYIGCGVDBTC-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=NC=CN1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results