CAS RN 33786-89-9
IUPAC Name:
5-chlorobenzene-1,3-diamine
Synonyms:
5-chlorobenzene-1,3-diamine
Molecular Weight (g/mol):
142.59
Molecular Formula:
C6H7ClN2
InChi Key:
VZNUCJOYPXKLTA-UHFFFAOYSA-N
SMILES:
NC1=CC(Cl)=CC(N)=C1
1
–
1
of
1
results
1
5-Chloro-m-phenylenediamine, 97%
CAS: 33786-89-9 Molecular Formula: C6H7ClN2 Molecular Weight (g/mol): 142.586 MDL Number: MFCD00014775 InChI Key: VZNUCJOYPXKLTA-UHFFFAOYSA-N Synonym: 5-chloro-m-phenylenediamine,3,5-diaminochlorobenzene,5-chloro-1,3-benzenediamine,1,3-benzenediamine, 5-chloro,5-chloro-1,3-phenylenediamine,ccris 2886,5-amino-3-chloroaniline,acmc-20am42,2-13-00-00029 beilstein handbook reference,5-chloro-1,3-diaminobenzene PubChem CID: 73335 IUPAC Name: 5-chlorobenzene-1,3-diamine SMILES: C1=C(C=C(C=C1N)Cl)N