accessibility menu, dialog, popup

UserName

FFFFFFFFFFFNFFFFFFFFFFFFFFFFFFFFFF

CAS RN 338-84-1

IUPAC Name: tris(1,1,2,2,3,3,4,4,5,5,5-undecafluoropentyl)amine
Synonyms: perfluorotripentylamine
Molecular Weight (g/mol): 821.12
Molecular Formula: C15F33N
InChi Key: AQZYBQIAUSKCCS-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)N(C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Boiling Point 
  • (2)
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results