accessibility menu, dialog, popup

UserName

H3CSOONNHN

CAS RN 338393-13-8

IUPAC Name: 5-[(4-methylphenyl)methanesulfonyl]-1H-1,2,4-triazole
Synonyms: 3-[(4-methylphenyl)methanesulfonyl]-2H-1,2,4-triazole
Molecular Weight (g/mol): 237.28
Molecular Formula: C10H11N3O2S
InChi Key: YYQTWXHTIYSIRV-UHFFFAOYSA-N
SMILES: CC1=CC=C(CS(=O)(=O)C2=NC=NN2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results