accessibility menu, dialog, popup

UserName

H3CSiCH3H3CSSiCH3CH3H3C

CAS RN 3385-94-2

IUPAC Name: trimethyl[(trimethylsilyl)sulfanyl]silane
Synonyms: bis(trimethylsilyl)sulfide
Molecular Weight (g/mol): 178.44
Molecular Formula: C6H18SSi2
InChi Key: RLECCBFNWDXKPK-UHFFFAOYSA-N
SMILES: C[Si](C)(C)S[Si](C)(C)C
Boiling Point 
  • (4)
  • (3)
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (3)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (4)
  • (4)
  • (3)
  • (4)
  • (3)
  • (7)
  • (3)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)