accessibility menu, dialog, popup

UserName

NNCuNNNNNNNNNNSSSSNNNNCH3H3CCH3H3CCH3H3CCH3H3CH3CCH3CH3CH3H3CCH3CH3CH3ClClClCl

CAS RN 33864-99-2

IUPAC Name: alcian blue stain
Synonyms: alcian blue stain
Molecular Weight (g/mol): 1298.86
Molecular Formula: C56H68Cl4CuN16S4
InChi Key: KDXHLJMVLXJXCW-UHFFFAOYSA-J
SMILES: [Cl-].[Cl-].[Cl-].[Cl-].CN(C)C(SCC1=CC2=C3\N=C4/N=C(\N=C5/N6[Cu]N3C(\N=C3/N=C(\N=C6\C6=CC(CSC(N(C)C)=[N+](C)C)=CC=C56)C5=CC=C(CSC(N(C)C)=[N+](C)C)C=C35)=C2C=C1)C1=CC=C(CSC(N(C)C)=[N+](C)C)C=C41)=[N+](C)C
Color 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)