accessibility menu, dialog, popup

UserName

H3CBrOO

CAS RN 33884-43-4

IUPAC Name: 2-(1-bromoethyl)-1,3-dioxane
Synonyms: 2-(1-bromoethyl)-1,3-dioxane
Molecular Weight (g/mol): 195.06
Molecular Formula: C6H11BrO2
InChi Key: KXMZOKKPQZRPRN-UHFFFAOYNA-N
SMILES: CC(Br)C1OCCCO1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (1)
  • (3)
  • (3)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (4)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)