CAS RN 3392-12-9
CAS RN 3392-12-9
IUPAC Name:
2,5-dioxopyrrolidin-1-yl 2-{[(tert-butoxy)carbonyl]amino}-3-methylbutanoate
Synonyms:
2,5-dioxopyrrolidin-1-yl 2-[(tert-butoxycarbonyl)amino]-3-methylbutanoate
Molecular Weight (g/mol):
314.34
Molecular Formula:
C14H22N2O6
InChi Key:
POBDBYGSGKMZPH-UHFFFAOYNA-N
SMILES:
CC(C)C(NC(=O)OC(C)(C)C)C(=O)ON1C(=O)CCC1=O