accessibility menu, dialog, popup

UserName

CAS RN 3392-97-0

CH3OOCH3O

CAS RN 3392-97-0

IUPAC Name: 2,6-dimethoxybenzaldehyde
Synonyms: 2,6-dimethoxybenzaldehyde
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: WXSGQHKHUYTJNB-UHFFFAOYSA-N
SMILES: COC1=CC=CC(OC)=C1C=O
Boiling Point 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)