accessibility menu, dialog, popup

UserName

ClON

CAS RN 33948-19-5

IUPAC Name: 2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaene-2-carbonyl chloride
Synonyms: 2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaene-2-carbonyl chloride
Molecular Weight (g/mol): 257.72
Molecular Formula: C15H12ClNO
InChi Key: COHHZMJBMIHLGF-UHFFFAOYSA-N
SMILES: ClC(=O)N1C2=CC=CC=C2CCC2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results