accessibility menu, dialog, popup

UserName

CAS RN 342385-52-8

HO(S)OH(R)OH(S)OHOOO

CAS RN 342385-52-8

IUPAC Name: (3S,4R,5S)-3,4,5,6-tetrahydroxy-2-oxohexanoate
Synonyms: 2-keto-L-gulonate L-xylo-hex-2-ulosonate 2-oxo-L-gulonate L-sorbosonate 2-dehydro-L-idonate
Molecular Weight (g/mol): 193.13
Molecular Formula: C6H9O7
InChi Key: VBUYCZFBVCCYFD-NUNKFHFFSA-M
SMILES: OC[C@H](O)[C@@H](O)[C@H](O)C(=O)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results