accessibility menu, dialog, popup

UserName

NSNNCO

CAS RN 342411-14-7

IUPAC Name: 4-isocyanato-2,1,3-benzothiadiazole
Synonyms: 4-isocyanato-2,1,3-benzothiadiazole
Molecular Weight (g/mol): 177.18
Molecular Formula: C7H3N3OS
InChi Key: HPZZFYLZFYDNMA-UHFFFAOYSA-N
SMILES: O=C=NC1=CC=CC2=NSN=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results