accessibility menu, dialog, popup

UserName

CH3H3C(S)NH3CH3H3CNHO

CAS RN 34272-83-8

IUPAC Name: (3S)-1-hydroxy-2,2,5,5-tetramethylpyrrolidin-3-aminium
Synonyms: (3S)-1-hydroxy-2,2,5,5-tetramethylpyrrolidin-3-aminium
Molecular Weight (g/mol): 159.25
Molecular Formula: C8H19N2O
InChi Key: USGBLEMQARVRDC-LURJTMIESA-O
SMILES: CC1(C)C[C@H]([NH3+])C(C)(C)N1O
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)