accessibility menu, dialog, popup

UserName

CAS RN 34298-75-4

IUPAC Name: (hex-4-yn-1-yl)benzene
Synonyms: hex-4-yn-1-ylbenzene
Molecular Weight (g/mol): 158.24
Molecular Formula: C12H14
InChi Key: BYQGPSNMZZGRQP-UHFFFAOYSA-N
SMILES: CC#CCCCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results