accessibility menu, dialog, popup

UserName

CAS RN 344-25-2

HOOHN

CAS RN 344-25-2

IUPAC Name: pyrrolidine-2-carboxylic acid
Synonyms: proline prolin Hpro proline DL-form
Molecular Weight (g/mol): 115.13
Molecular Formula: C5H9NO2
InChi Key: ONIBWKKTOPOVIA-UHFFFAOYNA-N
SMILES: OC(=O)C1CCCN1
Molecular Weight (g/mol) 
  • (3)
  • (7)
Quantity 
  • (3)
  • (1)
  • (2)
  • (4)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (7)
  • (3)
  • (1)
  • (2)
  • (4)
  • (3)
  • (3)
  • (1)