CAS RN 34454-97-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,1,2,2,3,3,4,4,4-Nonafluoro-N-(2-hydroxyethyl)-N-methyl-1-butanesulfonamide, TRC
CAS: 34454-97-2 Molecular Formula: C7 H8 F9 N O3 S Molecular Weight (g/mol): 357.19 Synonym: 2-[N-(Perfluorobutylsulfonyl)-N-methylamino]ethanol,N-(2-Hydroxyethyl)-N-methyl-perfluorobutane-1-sulfonamide,1,1,2,2,3,3,4,4,4-Nonafluoro-N-(2-hydroxyethyl)-N-methyl-1-butanesulfonamide IUPAC Name: 1,1,2,2,3,3,4,4,4-nonafluoro-N-(2-hydroxyethyl)-N-methyl-butane-1-sulfonamide SMILES: CN(CCO)S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F