CAS RN 34454-99-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,1,2,2,3,3,4,4,4-Nonafluoro-N-(2-hydroxyethyl)-1-butanesulfonamide, TRC
CAS: 34454-99-4 Molecular Formula: C6H6F9NO3S Molecular Weight (g/mol): 343.17 Synonym: N(2-Hydroxyethyl)perfluorobutanesulfonamide,N-(2-Hydroxyethyl)-1,1,2,2,3,3,4,4,4-nonafluorobutanesulfonamide; IUPAC Name: 1,1,2,2,3,3,4,4,4-nonafluoro-N-(2-hydroxyethyl)butane-1-sulfonamide SMILES: OCCNS(=O)(C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)=O