accessibility menu, dialog, popup

UserName

CH3ONCS

CAS RN 3460-49-9

IUPAC Name: 1-ethoxy-4-isothiocyanatobenzene
Synonyms: 1-ethoxy-4-isothiocyanatobenzene
Molecular Weight (g/mol): 179.24
Molecular Formula: C9H9NOS
InChi Key: REEKOMRZYJXXNR-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(C=C1)N=C=S
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results