accessibility menu, dialog, popup

UserName

CAS RN 34666-17-6

CH3CH3H3COOH3C

CAS RN 34666-17-6

IUPAC Name: ethyl 2,2-diethylbutanoate
Synonyms: ethyl 2,2-diethylbutanoate
Molecular Weight (g/mol): 172.27
Molecular Formula: C10H20O2
InChi Key: LXVSPQCOBPFDOO-UHFFFAOYSA-N
SMILES: CCOC(=O)C(CC)(CC)CC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results