accessibility menu, dialog, popup

UserName

CAS RN 34761-09-6

H3COOSH2N

CAS RN 34761-09-6

IUPAC Name: ethyl 3-amino-1-benzothiophene-2-carboxylate
Synonyms: ethyl 3-amino-1-benzothiophene-2-carboxylate
Molecular Weight (g/mol): 221.27
Molecular Formula: C11H11NO2S
InChi Key: XZSFZYVMRZLUOJ-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(N)C2=CC=CC=C2S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results