CAS RN 347840-08-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
(±)-2-Methyl-d3-succinic-2,3,3-d3 Acid, TRC
CAS: 347840-08-8 Molecular Formula: C5 D6 H2 O4 Molecular Weight (g/mol): 138.152 Synonym: Butanedioic-d3 acid, methyl-d3- (9CI),Methyl-d3-butanedioic-d3 acid,(±)-2-Methyl-d3-succinic-2,3,3-d3 Acid,Methylsuccinic Acid-D6,2-(Methyl-d3)butanedioic-2,3,3-d3 acid,2-(Methyl-d3)succinic-2,3,3-d3 acid IUPAC Name: 2,2,3-trideuterio-3-(trideuteriomethyl)butanedioic acid SMILES: [2H]C([2H])([2H])C([2H])(C(=O)O)C([2H])([2H])C(=O)O
2
(±)-2-Methyl-d3-succinic-2,3,3-d3 Acid, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.