accessibility menu, dialog, popup

UserName

NaH3CNCNH3CSOOOSOOO

CAS RN 3486-30-4

IUPAC Name: sodium bis({4-[benzyl(ethyl)amino]phenyl})(2,4-disulfonatophenyl)methylium
Synonyms: sodium bis({4-[benzyl(ethyl)amino]phenyl})(2,4-disulfonatophenyl)methylium
Molecular Weight (g/mol): 690.8
Molecular Formula: C37H35N2NaO6S2
InChi Key: FTUYQIPAPWPHNC-UHFFFAOYSA-M
SMILES: [Na+].CCN(CC1=CC=CC=C1)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC=C1)C1=CC=C(C=C1S([O-])(=O)=O)S([O-])(=O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results