CAS RN 34901-14-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Poly(ethylene glycol) bis(3-aminopropyl) terminated
CAS: 34901-14-9 Molecular Formula: (C2H4O)nC6H16N2O