accessibility menu, dialog, popup

UserName

ONOClN

CAS RN 351003-20-8

IUPAC Name: 3-(6-chloro-3-oxo-3,4-dihydro-2H-1,4-benzoxazin-4-yl)propanenitrile
Synonyms: 3-(6-chloro-3-oxo-2H-1,4-benzoxazin-4-yl)propanenitrile
Molecular Weight (g/mol): 236.66
Molecular Formula: C11H9ClN2O2
InChi Key: WLSJNBIUCRGWQJ-UHFFFAOYSA-N
SMILES: ClC1=CC2=C(OCC(=O)N2CCC#N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results