accessibility menu, dialog, popup

UserName

FFFN

CAS RN 3512-18-3

IUPAC Name: 2,3,6-trifluoropyridine
Synonyms: 2,3,6-trifluoropyridine
Molecular Weight (g/mol): 133.07
Molecular Formula: C5H2F3N
InChi Key: HRLIANGJWPJFKW-UHFFFAOYSA-N
SMILES: FC1=CC=C(F)C(F)=N1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results