CAS RN 3513-81-3
CAS RN 3513-81-3
IUPAC Name:
2-methylidenepropane-1,3-diol
Synonyms:
2-methylidenepropane-1,3-diol
Molecular Weight (g/mol):
88.11
Molecular Formula:
C4H8O2
InChi Key:
JFFYKITVXPZLQS-UHFFFAOYSA-N
SMILES:
OCC(=C)CO
1
–
1
of
1
results
1
2-Methylene-1,3-propanediol, 97%
CAS: 3513-81-3 Molecular Formula: C4H8O2 Molecular Weight (g/mol): 88.11 MDL Number: MFCD00075162 InChI Key: JFFYKITVXPZLQS-UHFFFAOYSA-N Synonym: 2-methylene-1,3-propanediol,2-methylenepropane-1,3-diol,1,3-propanediol, 2-methylene,1,3-dihydroxy-2-methylenepropane,1,3-dihydroxy-2-methylidenepropane,dimethylol ethylene,2-methylene1,3-propanediol,2-methylenepropan-1,3-diol,ksc573o4t,1,3-propanediol,2-methylene PubChem CID: 77048 IUPAC Name: 2-methylidenepropane-1,3-diol SMILES: C=C(CO)CO