accessibility menu, dialog, popup

UserName

H3N(R)(R)NH3

CAS RN 35132-20-8

IUPAC Name: (1R,2R)-1,2-diphenylethane-1,2-bis(aminium)
Synonyms: diphenylethylenediamine
Molecular Weight (g/mol): 214.31
Molecular Formula: C14H18N2
InChi Key: PONXTPCRRASWKW-ZIAGYGMSSA-P
SMILES: [NH3+][C@@H]([C@H]([NH3+])C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (6)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (4)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (2)
  • (6)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (2)