accessibility menu, dialog, popup

UserName

BFFFFRh

CAS RN 35138-22-8

IUPAC Name: bis(cycloocta-1,5-diene) tetrafluoroboranuide rhodium
Synonyms: bis(1,5-cyclooctadiene) rhodium tetrafluoroborate
Molecular Weight (g/mol): 406.08
Molecular Formula: C16H24BF4Rh
InChi Key: LYXHWHHENVLYCN-UHFFFAOYSA-N
SMILES: [Rh].F[B-](F)(F)F.C1CC=CCCC=C1.C1CC=CCCC=C1
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results