CAS RN 35179-98-7
CAS RN 35179-98-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Chloromethyl Ethyl Carbonate, TRC
CAS: 35179-98-7 Molecular Formula: C4H7ClO3 Molecular Weight (g/mol): 138.55 Synonym: Carbonic Acid Chloromethyl Ethyl Ester IUPAC Name: chloromethyl ethyl carbonate SMILES: CCOC(=O)OCCl